Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 18 - 19 | 2376 | 89 | |
GenBank mRNA | AK052404.1 | - | 18 - 19 | 2357 | 89 | |
AK054305.1 | - | 18 - 19 | 2358 | 89 | ||
AK077847.1 | - | 18 - 19 | 2377 | 89 | ||
AK085014.1 | - | 1 - 2 | 2369 | 89 | ||
AK090134.1 | - | 18 - 19 | 2358 | 89 | ||
AK161161.1 | - | 5 - 6 | 484 | 89 | ||
BC139002.1 | - | 18 - 19 | 2357 | 8862024 | 89 | |
BC145492.1 | - | 18 - 19 | 2357 | 9054055 | 89 | |
X80169.1 | - | 18 - 19 | 2361 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |