Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045484.1 | NP_001038949.1 | 4 - 5 | 481 | 96 | |
NM_008578.2 | NP_032604.2 | 4 - 5 | 481 | 96 | ||
GenBank mRNA | BC045147.1 | - | 4 - 5 | 481 | 6307224 | 96 |
CO811773.1 | - | 148 | 96 | |||
D87833.1 | - | 4 - 5 | 96 | |||
D87834.1 | - | 4 - 5 | 96 | |||
D87835.1 | - | 4 - 5 | 96 | |||
D87836.1 | - | 4 - 5 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |