Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163630.1 | NP_001157102.1 | 7 - 8 | 1528 | 96 | |
NM_001163631.1 | NP_001157103.1 | 7 - 8 | 1396 | 96 | ||
NM_001163632.1 | NP_001157104.1 | 7 - 8 | 1369 | 96 | ||
NM_009122.2 | NP_033148.2 | 7 - 8 | 1419 | 96 | ||
GenBank mRNA | AK037740.1 | - | 7 - 8 | 1420 | 96 | |
AK088563.1 | - | 7 - 8 | 1395 | 96 | ||
AK134040.1 | - | 7 - 8 | 1399 | 96 | ||
AK158518.1 | - | 7 - 8 | 1529 | 96 | ||
BC011132.1 | - | 7 - 8 | 1279 | 4164993 | 96 | |
CT010371.1 | - | 6 - 7 | 1207 | 96 | ||
U05252.1 | - | 7 - 8 | 1405 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |