Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164790.1 | NP_001158262.1 | 7 - 8 | 1201 | 77 | |
NM_009130.3 | NP_033156.1 | 7 - 8 | 1201 | 77 | ||
GenBank mRNA | AK133655.1 | - | 7 - 8 | 1103 | 77 | |
AK139140.1 | - | 7 - 8 | 1141 | 77 | ||
AK158315.1 | - | 7 - 8 | 1074 | 77 | ||
AK164089.1 | - | 7 - 8 | 1144 | 77 | ||
BC024785.1 | - | 7 - 8 | 1160 | 5362975 | 77 | |
U02982.1 | - | 7 - 8 | 1165 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |