Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009130.3 | NP_033156.1 | 9 - 10 | 1416 | 99 | |
GenBank mRNA | AK133655.1 | - | 9 - 10 | 1318 | 99 | |
AK139140.1 | - | 9 - 10 | 1356 | 99 | ||
AK148498.1 | - | 9 - 10 | 1400 | 99 | ||
U02982.1 | - | 9 - 10 | 1380 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |