Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009162.3 | NP_033188.3 | 2 - 3 | 327 | 77 | |
GenBank mRNA | AK003049.1 | - | 2 - 3 | 266 | 77 | |
AK005331.1 | - | 2 - 3 | 276 | 77 | ||
AK011938.1 | - | 2 - 3 | 258 | 77 | ||
AK017481.1 | - | 2 - 3 | 380 | 77 | ||
AK019337.1 | - | 2 - 3 | 328 | 77 | ||
AK031856.1 | - | 2 - 3 | 261 | 77 | ||
AK161556.1 | - | 2 - 3 | 266 | 77 | ||
AK161579.1 | - | 2 - 3 | 268 | 77 | ||
BC029021.1 | - | 2 - 3 | 309 | 5361810 | 77 | |
X15830.1 | - | 2 - 3 | 305 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |