Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009167.3 | NP_033193.2 | 7 - 8 | 608 | 65 | |
GenBank mRNA | BC103611.1 | - | 7 - 8 | 608 | 40046304 | 65 |
BC103612.1 | - | 7 - 8 | 608 | 40046306 | 65 | |
BC105644.1 | - | 7 - 8 | 608 | 40046301 | 65 | |
BC105645.1 | - | 7 - 8 | 608 | 40046303 | 65 | |
U46854.1 | - | 7 - 8 | 608 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |