Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164639.1 | NP_001158111.1 | 1 - 2 | 322 | 98 | |
NM_009289.3 | NP_033315.2 | 1 - 2 | 322 | 98 | ||
GenBank mRNA | AF039574.1 | - | 1 - 2 | 327 | 98 | |
AF112855.1 | - | 1 - 2 | 211 | 98 | ||
AK029491.1 | - | 1 - 2 | 189 | 98 | ||
AK122218.1 | - | 1 - 2 | 663 | 98 | ||
BC131674.1 | - | 1 - 2 | 223 | 40110915 | 98 | |
BC131675.1 | - | 1 - 2 | 223 | 40110921 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |