Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009796.2 | NP_033926.1 | 16 - 17 | 2025 | 74 | |
GenBank mRNA | AB028640.1 | - | 16 - 17 | 2025 | 74 | |
AJ012475.1 | - | 16 - 17 | 1838 | 74 | ||
AK154277.1 | - | 16 - 17 | 2111 | 74 | ||
AK169334.1 | - | 16 - 17 | 3070 | 74 | ||
BC095980.1 | - | 16 - 17 | 3040 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |