Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141737.1 | - | 10 - 11 | 1140 | 122 | |
AK168287.1 | - | 10 - 11 | 1135 | 122 | ||
BC004669.1 | - | 9 - 10 | 844 | 3498846 | 122 | |
L16926.1 | - | 9 - 10 | 1322 | 122 | ||
U15562.1 | - | 10 - 11 | 1106 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |