Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009867.2 | NP_033997.1 | 10 - 11 | 1761 | 56 | |
GenBank mRNA | D14888.1 | - | 10 - 11 | 1671 | 56 | |
X69966.1 | - | 10 - 11 | 1617 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |