Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009879.3 | NP_034009.2 | 13 - 14 | 1563 | 111 | |
GenBank mRNA | AF354757.1 | - | 13 - 14 | 1563 | 111 | |
BC055721.1 | - | 13 - 14 | 1520 | 6810406 | 111 | |
Y10495.2 | - | 13 - 14 | 1563 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |