Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009923.2 | NP_034053.2 | 1 - 2 | 123 | 89 | |
GenBank mRNA | AK050628.1 | - | 1 - 2 | 88 | 89 | |
AK087415.1 | - | 1 - 2 | 89 | 89 | ||
AK154654.1 | - | 1 - 2 | 147 | 89 | ||
AK158318.1 | - | 1 - 2 | 117 | 89 | ||
AK172318.1 | - | 1 - 2 | 88 | 89 | ||
AK172514.1 | - | 1 - 2 | 88 | 89 | ||
BC005544.1 | - | 1 - 2 | 76 | 3585034 | 89 | |
BY333311.1 | - | 1 - 2 | 124 | 89 | ||
M58045.1 | - | 1 - 2 | 46 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |