Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009996.3 | NP_034126.1 | 4 - 5 | 1005 | 70 | |
GenBank mRNA | AK143984.1 | - | 4 - 5 | 1013 | 70 | |
AK159109.1 | - | 4 - 5 | 1013 | 70 | ||
AK159481.1 | - | 4 - 5 | 1013 | 70 | ||
AK159527.1 | - | 4 - 5 | 1013 | 70 | ||
AK159852.1 | - | 4 - 5 | 1013 | 70 | ||
AK159954.1 | - | 4 - 5 | 1013 | 70 | ||
AK159976.1 | - | 4 - 5 | 1013 | 70 | ||
AK164697.1 | - | 4 - 5 | 1013 | 70 | ||
BC109139.1 | - | 4 - 5 | 997 | 40039500 | 70 | |
BC109140.1 | - | 4 - 5 | 997 | 40039502 | 70 | |
D49438.1 | - | 4 - 5 | 943 | 70 | ||
D89669.1 | - | 4 - 5 | 1005 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |