Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010008.4 | NP_034138.3 | 1 - 2 | 408 | 116 | |
GenBank mRNA | AK033614.1 | - | 1 - 2 | 411 | 116 | |
AK045421.1 | - | 1 - 2 | 398 | 116 | ||
AK078893.1 | - | 1 - 2 | 234 | 116 | ||
AK084025.1 | - | 1 - 2 | 418 | 116 | ||
AK165663.1 | - | 1 - 2 | 253 | 116 | ||
AK180579.1 | - | 1 - 2 | 97 | 116 | ||
AK186838.1 | - | 1 - 2 | 97 | 116 | ||
AK210777.1 | - | 1 - 2 | 194 | 116 | ||
BC050832.1 | - | 1 - 2 | 402 | 6514145 | 116 | |
U62295.1 | - | 1 - 2 | 455 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection P450 Mus musculus 1700 Inventoried Probe spans exons |