Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010247.2 | NP_034377.2 | 4 - 5 | 449 | 123 | |
GenBank mRNA | AB010282.1 | - | 4 - 5 | 390 | 123 | |
AF483500.1 | - | 3 - 4 | 329 | 123 | ||
AF483501.1 | - | 3 - 4 | 329 | 123 | ||
AK143570.1 | - | 4 - 5 | 424 | 123 | ||
AK146392.1 | - | 4 - 5 | 368 | 123 | ||
AK146394.1 | - | 4 - 5 | 368 | 123 | ||
AK146396.1 | - | 4 - 5 | 416 | 123 | ||
BC031422.1 | - | 4 - 5 | 398 | 3669715 | 123 | |
BC051085.1 | - | 4 - 5 | 1627 | 123 | ||
M38700.1 | - | 3 - 4 | 291 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |