Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288586.1 | NP_001275515.1 | 4 - 5 | 494 | 97 | |
NM_010786.4 | NP_034916.1 | 5 - 6 | 579 | 97 | ||
GenBank mRNA | AK004719.1 | - | 5 - 6 | 538 | 97 | |
AK004881.1 | - | 5 - 6 | 506 | 97 | ||
AK088638.1 | - | 5 - 6 | 509 | 97 | ||
AK150530.1 | - | 4 - 5 | 367 | 97 | ||
AK152685.1 | - | 4 - 5 | 367 | 97 | ||
AK160069.1 | - | 4 - 5 | 370 | 97 | ||
AK168690.1 | - | 5 - 6 | 579 | 97 | ||
BC050902.1 | - | 5 - 6 | 495 | 6415937 | 97 | |
BC092270.1 | - | 5 - 6 | 532 | 4018419 | 97 | |
CN703560.1 | - | 4 - 5 | 436 | 97 | ||
U47934.1 | - | 3 - 4 | 285 | 97 | ||
X58876.1 | - | 5 - 6 | 486 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |