Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhlha15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010800.4 | NP_034930.1 | 1 - 2 | 68 | 115 | |
GenBank mRNA | AF049660.1 | - | 1 - 2 | 68 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |