Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010808.3 | NP_034938.3 | 3 - 4 | 566 | 63 | |
GenBank mRNA | AB021226.1 | - | 3 - 4 | 511 | 63 | |
AJ010262.1 | - | 3 - 4 | 512 | 63 | ||
AY622976.1 | - | 2 - 3 | 155 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |