Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011176.4 | NP_035306.2 | 11 - 12 | 1529 | 74 | |
GenBank mRNA | AF042822.3 | - | 11 - 12 | 1418 | 74 | |
AK052738.1 | - | 11 - 12 | 1529 | 74 | ||
AK166745.1 | - | 11 - 12 | 1520 | 74 | ||
BC005496.1 | - | 11 - 12 | 1528 | 3488059 | 74 | |
BC008514.1 | - | 11 - 12 | 1528 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |