Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277938.1 | NP_001264867.1 | 6 - 7 | 68 | ||
NM_001277939.1 | NP_001264868.1 | 6 - 7 | 68 | |||
NM_001277941.1 | NP_001264870.1 | 5 - 6 | 68 | |||
NM_011235.4 | NP_035365.1 | 6 - 7 | 68 | |||
NR_102717.1 | 6 - 7 | 68 | ||||
NR_102718.1 | 6 - 7 | 68 | ||||
NR_102719.1 | 5 - 6 | 68 | ||||
NR_102720.1 | 5 - 6 | 68 | ||||
GenBank mRNA | AB007040.1 | - | 6 - 7 | 68 | ||
AB052828.1 | - | 6 - 7 | 748 | 68 | ||
AB052829.1 | - | 6 - 7 | 748 | 68 | ||
AB052831.1 | - | 5 - 6 | 629 | 68 | ||
AB052832.1 | - | 5 - 6 | 629 | 68 | ||
AB052833.1 | - | 5 - 6 | 613 | 68 | ||
AB052834.1 | - | 6 - 7 | 771 | 68 | ||
AF034955.1 | - | 6 - 7 | 68 | |||
AK036272.1 | - | 5 - 6 | 68 | |||
AK138553.1 | - | 6 - 7 | 68 | |||
AK147759.1 | - | 6 - 7 | 68 | |||
BC049136.1 | - | 6 - 7 | 6832014 | 68 | ||
CF746344.1 | - | 2 - 3 | 68 | |||
EU627687.1 | - | 6 - 7 | 68 | |||
EU627688.1 | - | 6 - 7 | 68 | |||
Y15570.1 | - | 6 - 7 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |