Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NM_001277938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NM_001277941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NM_001277942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NM_011235.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NR_102717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NR_102718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NR_102719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
NR_102720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030930.mm.1 |
TC1100003372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NM_001277938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NM_001277939.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NM_001277941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NM_011235.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NR_102718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NR_102719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
NR_102720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030935.mm.1 |
TC1100003372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NM_001277938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NM_001277941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NM_001277942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NM_011235.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NR_102717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NR_102718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NR_102719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
NR_102720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030929.mm.1 |
TC1100003372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NM_001277938.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NM_001277941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NM_001277942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NM_011235.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NR_102717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NR_102718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NR_102719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
NR_102720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030928.mm.1 |
TC1100003372.mm.1 |
MTA TC |