Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011237.2 | NP_035367.1 | 3 - 4 | 318 | 60 | |
GenBank mRNA | AF045663.1 | - | 3 - 4 | 318 | 60 | |
AK134293.1 | - | 3 - 4 | 331 | 60 | ||
AK138973.1 | - | 3 - 4 | 323 | 60 | ||
AK165833.1 | - | 3 - 4 | 353 | 60 | ||
BC082556.1 | - | 3 - 4 | 257 | 30538342 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |