Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081459.2 | NP_001074928.1 | 3 - 4 | 1636 | 67 | |
NM_001289538.1 | NP_001276467.1 | 3 - 4 | 1636 | 67 | ||
NM_001289539.1 | NP_001276468.1 | 3 - 4 | 1259 | 67 | ||
NM_001289540.1 | NP_001276469.1 | 4 - 5 | 1396 | 67 | ||
NM_001289541.1 | NP_001276470.1 | 3 - 4 | 1259 | 67 | ||
NM_001289542.1 | NP_001276471.1 | 3 - 4 | 1636 | 67 | ||
NM_011363.3 | NP_035493.2 | 3 - 4 | 1636 | 67 | ||
NR_110346.1 | 2 - 3 | 1124 | 67 | |||
GenBank mRNA | AF020526.1 | - | 3 - 4 | 1330 | 67 | |
AF074329.1 | - | 2 - 3 | 1041 | 67 | ||
AF380422.1 | - | 3 - 4 | 1330 | 67 | ||
AF421138.1 | - | 3 - 4 | 1330 | 67 | ||
AF421139.1 | - | 3 - 4 | 1330 | 67 | ||
AK148085.1 | - | 4 - 5 | 1363 | 67 | ||
AK168439.1 | - | 3 - 4 | 1231 | 67 | ||
AK170444.1 | - | 3 - 4 | 1610 | 67 | ||
AK171328.1 | - | 3 - 4 | 1644 | 67 | ||
AK173145.1 | - | 2 - 3 | 783 | 67 | ||
BC011422.1 | - | 2 - 3 | 1098 | 3156603 | 67 | |
BC051978.1 | - | 3 - 4 | 1179 | 6401205 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |