Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 9 - 10 | 1699 | 88 | |
NM_001110351.1 | NP_001103821.1 | 10 - 11 | 1576 | 88 | ||
NM_011378.2 | NP_035508.2 | 10 - 11 | 1807 | 88 | ||
GenBank mRNA | AK032209.1 | - | 9 - 10 | 1567 | 88 | |
AK136347.1 | - | 4 - 5 | 501 | 88 | ||
AK220292.1 | - | 9 - 10 | 1676 | 88 | ||
BC052716.1 | - | 9 - 10 | 1586 | 6837144 | 88 | |
BC053385.1 | - | 9 - 10 | 1569 | 88 | ||
BC078454.1 | - | 9 - 10 | 1560 | 88 | ||
L36831.1 | - | 9 - 10 | 1490 | 88 | ||
L38620.2 | - | 8 - 9 | 1466 | 88 | ||
L38621.2 | - | 8 - 9 | 1466 | 88 | ||
U22394.1 | - | 10 - 11 | 1815 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |