Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011499.3 | NP_035629.2 | 2 - 3 | 523 | 91 | |
GenBank mRNA | AF096285.1 | - | 3 - 4 | 335 | 91 | |
AK075804.1 | - | 2 - 3 | 481 | 91 | ||
AK077504.1 | - | 2 - 3 | 475 | 91 | ||
AK145971.1 | - | 2 - 3 | 475 | 91 | ||
AK147179.1 | - | 2 - 3 | 524 | 91 | ||
AK152283.1 | - | 2 - 3 | 373 | 91 | ||
AK154158.1 | - | 2 - 3 | 491 | 91 | ||
BC141076.1 | - | 2 - 3 | 474 | 9055688 | 91 | |
BC145342.1 | - | 2 - 3 | 474 | 9053898 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |