Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013729.3 | NP_038757.1 | 1 - 2 | 413 | 74 | |
GenBank mRNA | AB221547.1 | - | 1 - 2 | 395 | 74 | |
AF135063.1 | - | 1 - 2 | 391 | 74 | ||
AF154573.1 | - | 1 - 2 | 413 | 74 | ||
AF201959.1 | - | 1 - 2 | 356 | 74 | ||
BC099876.1 | - | 1 - 2 | 413 | 40040047 | 74 | |
BC099877.1 | - | 1 - 2 | 413 | 40040048 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |