Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013809.1 | NP_038837.1 | 3 - 4 | 495 | 79 | |
GenBank mRNA | AK014160.1 | - | 3 - 4 | 514 | 79 | |
BC046321.1 | - | 3 - 4 | 530 | 79 | ||
BC058222.1 | - | 3 - 4 | 500 | 6306453 | 79 | |
L81171.1 | - | 3 - 4 | 495 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Mus musculus Inventoried Probe spans exons |