Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013809.1 | NP_038837.1 | 4 - 5 | 650 | 93 | |
GenBank mRNA | AK014160.1 | - | 4 - 5 | 669 | 93 | |
BC046321.1 | - | 4 - 5 | 685 | 93 | ||
BC058222.1 | - | 4 - 5 | 655 | 6306453 | 93 | |
L81171.1 | - | 4 - 5 | 650 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |