Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177572.1 | NP_001171043.1 | 11 - 12 | 1339 | 60 | |
NM_015829.3 | NP_056644.1 | 11 - 12 | 1342 | 60 | ||
GenBank mRNA | AF164632.1 | - | 11 - 12 | 1287 | 60 | |
AK002829.1 | - | 11 - 12 | 1217 | 60 | ||
AK012670.1 | - | 11 - 12 | 1230 | 60 | ||
AK029020.1 | - | 11 - 12 | 1270 | 60 | ||
BC016571.1 | - | 5 - 6 | 476 | 3600909 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |