Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016695.3 | NP_057904.1 | 6 - 7 | 811 | 69 | |
GenBank mRNA | AF162685.1 | - | 6 - 7 | 798 | 69 | |
AK141422.1 | - | 5 - 6 | 809 | 69 | ||
AK161577.1 | - | 6 - 7 | 804 | 69 | ||
AK172253.1 | - | 6 - 7 | 796 | 69 | ||
AK188703.1 | - | 1 - 2 | 92 | 69 | ||
BC053026.1 | - | 6 - 7 | 811 | 5709543 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |