Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016717.3 | NP_057926.2 | 10 - 11 | 1130 | 65 | |
GenBank mRNA | AF175407.1 | - | 10 - 11 | 1082 | 65 | |
AK036750.1 | - | 10 - 11 | 1132 | 65 | ||
BC019879.1 | - | 10 - 11 | 1090 | 5123843 | 65 | |
BC021389.1 | - | 10 - 11 | 1114 | 5054691 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |