Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016769.4 | NP_058049.3 | 3 - 4 | 848 | 81 | |
GenBank mRNA | AB008192.1 | - | 3 - 4 | 557 | 81 | |
AF016189.1 | - | 3 - 4 | 824 | 81 | ||
AK041743.1 | - | 3 - 4 | 369 | 81 | ||
AK048626.1 | - | 3 - 4 | 848 | 81 | ||
AK083158.1 | - | 3 - 4 | 456 | 81 | ||
BC066850.1 | - | 3 - 4 | 534 | 30432720 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |