Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030955.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030955.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030955.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030955.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030955.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030956.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030956.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030956.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030956.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030956.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030957.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030957.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030957.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030957.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030957.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030959.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030959.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030959.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030959.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030959.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030960.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030960.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030960.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030960.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030960.mm.1 |
NM_016799.3 |
Refseq |