Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016856.3 | NP_058552.1 | 7 - 8 | 1040 | 108 | |
GenBank mRNA | AF012822.1 | - | 7 - 8 | 1026 | 108 | |
AK140869.1 | - | 7 - 8 | 1022 | 108 | ||
AK141325.1 | - | 7 - 8 | 1022 | 108 | ||
AK147746.1 | - | 7 - 8 | 1041 | 108 | ||
BC007163.1 | - | 5 - 6 | 698 | 108 | ||
BC013628.1 | - | 7 - 8 | 1015 | 3497854 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |