Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016856.3 | NP_058552.1 | 10 - 11 | 1640 | 61 | |
GenBank mRNA | AF012822.1 | - | 10 - 11 | 1626 | 61 | |
AK140869.1 | - | 10 - 11 | 1622 | 61 | ||
BC007163.1 | - | 8 - 9 | 1298 | 61 | ||
BC013628.1 | - | 10 - 11 | 1615 | 3497854 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |