Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252614.1 | NP_001239543.1 | 9 - 10 | 1433 | 110 | |
NM_001252615.1 | NP_001239544.1 | 3 - 4 | 507 | 110 | ||
NM_001252616.1 | NP_001239545.1 | 2 - 3 | 527 | 110 | ||
NM_016893.5 | NP_058589.2 | 8 - 9 | 1344 | 110 | ||
NR_045554.1 | 8 - 9 | 1763 | 110 | |||
GenBank mRNA | AB025198.1 | - | 5 - 6 | 1033 | 110 | |
AK048520.1 | - | 8 - 9 | 1482 | 110 | ||
AK051811.1 | - | 8 - 9 | 1301 | 110 | ||
AK134537.1 | - | 8 - 9 | 1437 | 110 | ||
AK211417.1 | - | 1 - 2 | 68 | 110 | ||
BC010666.1 | - | 9 - 10 | 1412 | 3594582 | 110 | |
EF591874.1 | - | 3 - 4 | 507 | 110 | ||
EF591875.1 | - | 2 - 3 | 527 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |