Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017474.2 | NP_059502.1 | 5 - 6 | 762 | 106 | |
GenBank mRNA | AB017156.1 | - | 5 - 6 | 745 | 106 | |
AK007466.1 | - | 5 - 6 | 759 | 106 | ||
AK008659.2 | - | 5 - 6 | 759 | 106 | ||
AK033560.1 | - | 763 | 106 | |||
AK136476.1 | - | 5 - 6 | 759 | 106 | ||
BC028343.1 | - | 5 - 6 | 752 | 4217548 | 106 | |
BC116318.2 | - | 5 - 6 | 745 | 40095991 | 106 | |
BC116319.2 | - | 5 - 6 | 745 | 40095992 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |