Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253700.1 | NP_001240629.1 | 6 - 7 | 642 | 62 | |
NM_018740.2 | NP_061210.2 | 8 - 9 | 1124 | 62 | ||
GenBank mRNA | AF004107.1 | - | 7 - 8 | 1043 | 62 | |
AK011130.1 | - | 6 - 7 | 936 | 62 | ||
AK013662.1 | - | 6 - 7 | 725 | 62 | ||
AK152582.1 | - | 8 - 9 | 1128 | 62 | ||
AK215451.1 | - | 1 - 2 | 40 | 62 | ||
BC003752.1 | - | 7 - 8 | 975 | 3489878 | 62 | |
CD741073.1 | - | 1 - 2 | 62 | |||
CD776986.1 | - | 1 - 2 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021055 | AF004107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021054 | BC003752.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021054 | AK215451.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021054 | CD776986.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021054 | AF004107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021054 | NM_018740.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650906 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692868 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576196 | AK003354.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576196 | BC002128.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576196 | NM_024206.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963988 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30340 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165955 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110653 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021054 | BC003752.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021054 | AK215451.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021054 | CD776986.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021054 | AF004107.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021054 | NM_018740.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898294 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898293 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165955 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07387268 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07387255 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110653 | - | - |
Whole Mouse Genome G4122A | A_51_P127777 | AF004107.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119990 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165955 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110653 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191734 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191748 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191742 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09225752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09694115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08976750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09443297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09031563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08981005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09043836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09694099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27898231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09653551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10387763 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387758 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387765 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387767 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387761 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387764 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387759 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387766 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387762 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431411_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10387757 | - | - |
Mouse Genome 430 2.0 Array | 1444136_at | - | - |
Mouse Genome 430 2.0 Array | 1431411_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1431411_a_at | - | - |
Mouse Genome 430 2.0 Array | 1444136_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54351_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54351_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100028349.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028349.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028349.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028349.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028349.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028368.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028368.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028368.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028368.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028368.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028363.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028363.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028363.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028363.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028363.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028364.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028364.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028364.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028364.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028364.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028355.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028355.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028355.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028355.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028355.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028361.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028361.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028361.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028361.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028361.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028373.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028373.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028373.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028376.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028376.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028376.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028360.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028360.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028360.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028356.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028356.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028356.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028356.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028356.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028350.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028350.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028350.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028350.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028350.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028374.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028374.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028374.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028372.mm.1 | NM_001253700.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028372.mm.1 | NM_001253700.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028372.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028372.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028372.mm.1 | TC1100003079.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028359.mm.1 | NM_018740.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028359.mm.1 | NM_018740.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028359.mm.1 | TC1100003079.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431411_a_at | - | - |
Mouse Expression Set 430 | 1431411_PM_a_at | - | - |
Murine Genome U74v2 Set | 160077_at | - | - |
Murine Genome U74v2 Set | 160077_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1431411_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1431411_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431411_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431411_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 511315 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1580093 | - | - |
MouseWG-6 v2 Expression BeadChip | 3830603 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1580093 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2570079 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3830603 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002570079 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003830603 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001580093 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235982 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238130 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214951 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214951 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238130 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1235982 | - | - |
mouse-6 v1.1 (Illumina) | scl0001572.1_0-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001363.1_5-S | - | - |
mouse-6 v1.1 (Illumina) | scl40005.7.1_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3830603 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1580093 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2570079 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl40005.7.1_7-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001363.1_5-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001572.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570079 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830603 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580093 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1214951 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1235982 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1238130 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2891573 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214951 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238130 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235982 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2891573 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001363.1_5-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001572.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40005.7.1_7-S | - | - |