Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018828.2 | NP_061298.1 | 14 - 15 | 2714 | 66 | |
GenBank mRNA | AK084018.1 | - | 14 - 15 | 2506 | 66 | |
AK129263.1 | - | 13 - 14 | 3585 | 66 | ||
U40750.2 | - | 14 - 15 | 2714 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |