Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205311.1 | NP_001192240.1 | 13 - 14 | 1697 | 64 | |
NM_019415.2 | NP_062288.2 | 13 - 14 | 1694 | 64 | ||
GenBank mRNA | AK052691.1 | - | 13 - 14 | 1698 | 64 | |
AK052755.1 | - | 13 - 14 | 1696 | 64 | ||
AK085496.1 | - | 13 - 14 | 1703 | 64 | ||
BC038612.1 | - | 13 - 14 | 1719 | 4237274 | 64 | |
U61085.1 | - | 13 - 14 | 1676 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |