Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205311.1 | NP_001192240.1 | 22 - 23 | 2691 | 66 | |
NM_019415.2 | NP_062288.2 | 22 - 23 | 2688 | 66 | ||
GenBank mRNA | AK052691.1 | - | 22 - 23 | 2692 | 66 | |
BC038612.1 | - | 22 - 23 | 2713 | 4237274 | 66 | |
U61085.1 | - | 22 - 23 | 2670 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |