Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166531.1 | NP_001160003.1 | 12 - 13 | 1525 | 114 | |
NM_001166532.1 | NP_001160004.1 | 11 - 12 | 1619 | 114 | ||
NM_019460.2 | NP_062333.1 | 11 - 12 | 1476 | 114 | ||
GenBank mRNA | AB032165.1 | - | 10 - 11 | 1321 | 114 | |
AK142330.1 | - | 11 - 12 | 1503 | 114 | ||
AK154826.1 | - | 11 - 12 | 1621 | 114 | ||
AK160573.1 | - | 12 - 13 | 1525 | 114 | ||
BC018217.1 | - | 11 - 12 | 1487 | 114 | ||
BC036972.1 | - | 11 - 12 | 1546 | 114 | ||
BC066188.1 | - | 11 - 12 | 1429 | 30476111 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |