Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167910.1 | NP_001161382.1 | 19 - 20 | 3015 | 85 | |
NM_019709.4 | NP_062683.3 | 20 - 21 | 3164 | 85 | ||
GenBank mRNA | AF094820.1 | - | 18 - 19 | 2583 | 85 | |
AK029048.1 | - | 19 - 20 | 2992 | 85 | ||
AK122202.1 | - | 19 - 20 | 2981 | 85 | ||
AK147336.1 | - | 18 - 19 | 5584 | 85 | ||
BC011533.1 | - | 19 - 20 | 2978 | 85 | ||
BC054837.1 | - | 18 - 19 | 2783 | 6490394 | 85 | |
BC057198.1 | - | 20 - 21 | 3130 | 5310414 | 85 | |
DQ913734.1 | - | 18 - 19 | 2646 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |