Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019721.2 | NP_062695.2 | 9 - 10 | 1649 | 87 | |
GenBank mRNA | AF135789.1 | - | 7 - 8 | 1212 | 87 | |
AK009492.1 | - | 6 - 7 | 710 | 87 | ||
AK035061.1 | - | 8 - 9 | 1467 | 87 | ||
AK085189.1 | - | 9 - 10 | 1636 | 87 | ||
BE979768.1 | - | 2 - 3 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |