Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163472.1 | NP_001156944.1 | 4 - 5 | 766 | 87 | |
NM_019823.4 | NP_062797.3 | 4 - 5 | 760 | 87 | ||
GenBank mRNA | AF221525.1 | - | 3 - 4 | 556 | 87 | |
AK090296.1 | - | 3 - 4 | 426 | 87 | ||
BC006853.1 | - | 3 - 4 | 1005 | 87 | ||
BC016256.1 | - | 4 - 5 | 668 | 4912394 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |