Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110807.1 | NP_001104277.1 | 8 - 9 | 1034 | 72 | |
GenBank mRNA | AJ289243.1 | - | 8 - 9 | 1042 | 72 | |
BC028751.1 | - | 8 - 9 | 1033 | 1314413 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |