Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021899.3 | NP_068699.1 | 3 - 4 | 1212 | 59 | |
GenBank mRNA | AB221666.1 | - | 2 - 3 | 411 | 59 | |
AF253052.1 | - | 3 - 4 | 1224 | 59 | ||
AK154574.1 | - | 3 - 4 | 1455 | 59 | ||
BC040395.1 | - | 4 - 5 | 842 | 4190087 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |