Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022317.3 | NP_071712.3 | 1 - 2 | 181 | 83 | |
GenBank mRNA | AF305211.1 | - | 1 - 2 | 114 | 83 | |
AK052784.1 | - | 1 - 2 | 182 | 83 | ||
AK145077.1 | - | 1 - 2 | 182 | 83 | ||
AK164752.1 | - | 1 - 2 | 182 | 83 | ||
BC010472.1 | - | 1 - 2 | 169 | 3586005 | 83 | |
BC013783.1 | - | 1 - 2 | 172 | 3600786 | 83 | |
BC037061.1 | - | 1 - 2 | 184 | 83 | ||
BY736278.1 | - | 1 - 2 | 182 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |