Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384314 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821898 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
503580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
151234 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
843399 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410616 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462307 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
678230 |
NM_146912.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56607 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157064 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P175782 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147613 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147631 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147590 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147629 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147602 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147600 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147626 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147623 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147625 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147620 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147630 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147605 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P175782 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08762164 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08762144 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
BC011228.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
BC094016.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AK005007.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AK018676.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452768 |
AB037540.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129724 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08762144 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P175782 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13439363 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13439349 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13439351 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13439350 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13439352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147525 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147575 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147592 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13439349 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147582 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147594 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147584 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147578 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147596 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147633 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147625 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147567 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147558 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147620 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147635 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147560 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147627 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147576 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147611 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13211440 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147579 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13450356 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147598 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147564 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147565 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13318781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147641 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147581 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147639 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147571 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147580 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147569 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147583 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13230390 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147626 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13450350 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147631 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147593 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147637 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147630 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147589 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147629 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147614 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147615 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147600 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13439371 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147586 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13125068 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147607 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147604 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147643 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147572 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13439364 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13450348 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147608 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147559 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147617 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13439374 |
- |
- |